Learn More
Analytical Sales & Services 2mL 96-Well Collection Plate, Square Top / Tapered V Bottom, Black
Catalog No. 5000804716
Encompass
Black 96-Well Plate 2mL Square Top Tapered V Bottom\n\n• Made in USA of domestically sourced and certified materials\n\nQty/Pkg: 10
Catalog No. 50-008-04716 Supplier Analytical Sales & Services Supplier No. 5962323BK
Black 96-Well Plate 2mL Square Top Tapered V Bottom\n\n• Made in USA of domestically sourced and certified materials\n\nQty/Pkg: 10