Learn More
Analytical Sales & Services 2mL, 96-Well Collection Plate with V-Bottom (pyramidal) Wells
Catalog No. 5000807132
Encompass
2mL 96 Well Collection Plate\n• Square Wells with V-Bottoms (Pyramid Shape)\n• Polypropylene\n\nQty/Pkg: 50
Catalog No. 50-008-07132 Supplier Analytical Sales & Services Supplier No. 968270
2mL 96 Well Collection Plate\n• Square Wells with V-Bottoms (Pyramid Shape)\n• Polypropylene\n\nQty/Pkg: 50