Learn More
Alkali Scientific CellPro™ 2.2ml 96-Well Deep Well Plate, V-Bottom, Square Well, 5/pk, 50/cs
SDP
Catalog No. 502388698
Encompass_Preferred
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).
Catalog No. 50-238-8698 Supplier Alkali Scientific Supplier No. MPD26
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).