Learn More
Analytical Sales & Services 2mL Amber Screw Vials and Caps - for Spectra-Physics Autoamplers
Catalog No. 5000804725
Encompass
Advantage™ Pre-Assembled Vial Kit. Includes: 8mm, 2mL Amber Glass Screw Vials and 8mm Black Phenolic Caps with Silicone/PTFE Liners For Spectra-Physics Autosamplers
Catalog No. 50-008-04725 Supplier Analytical Sales & Services Supplier No. 52004ACASE
Advantage™ Pre-Assembled Vial Kit. Includes: 8mm, 2mL Amber Glass Screw Vials and 8mm Black Phenolic Caps with Silicone/PTFE Liners For Spectra-Physics Autosamplers