Learn More
Bellco Glass, Inc. 2mL Aspirating Pipet
SDP
Catalog No. 502330326
Encompass_Preferred
Ideal for aspiration of media and sample waste removal without needing to measure the volumeSupplied without graduations or plugs2mL available in plastic/plastic wrap (Part No. 229262A) – Peel-to-open plastic/plastic wrappers are the ideal option for eliminating particulate; proven benefit over traditional paper/plastic wrapper Pack Description100 Individually Wrapped/BagCase Quantity400SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneMaximum Volume2mLGraduatedNoRNase/DNase FreeYes
Catalog No. 50-233-0326 Supplier Bellco Glass, Inc. Supplier No. 229262000
Ideal for aspiration of media and sample waste removal without needing to measure the volumeSupplied without graduations or plugs2mL available in plastic/plastic wrap (Part No. 229262A) – Peel-to-open plastic/plastic wrappers are the ideal option for eliminating particulate; proven benefit over traditional paper/plastic wrapper Pack Description100 Individually Wrapped/BagCase Quantity400SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneMaximum Volume2mLGraduatedNoRNase/DNase FreeYes