Learn More
Crystalgen Inc 2ML ASPIRATING PIPETTE 200/CS
SDP
Catalog No. NC2000104
Encompass_Preferred
Individually wrapped pipets, packaged in paper-plastic wrappers. Crystalgen Serological pipets consist of high quality virgin polystyrene with polyester filter. Manufactured in an ISO 9001:2008 certified facility.
Catalog No. NC2000104 Supplier Crystalgen Inc Supplier No. AS8002
Individually wrapped pipets, packaged in paper-plastic wrappers. Crystalgen Serological pipets consist of high quality virgin polystyrene with polyester filter. Manufactured in an ISO 9001:2008 certified facility.