Learn More
Ace Glass, Inc. 2ML SPEC GRAVITY BTL W/ STOPPR
SDP
Catalog No. 50971882
Encompass_Preferred
Guy-Lussac type 2mL specific gravity bottle with 7/12 stopper, unadjusted for calibration in the laboratory. When adjusted, the bottle is suitable for ASTM D 369.
Catalog No. 50-971-882 Supplier Ace Glass, Inc. Supplier No. 542006
Guy-Lussac type 2mL specific gravity bottle with 7/12 stopper, unadjusted for calibration in the laboratory. When adjusted, the bottle is suitable for ASTM D 369.