Learn More
Phenomenex Inc 2mL centrifugal tube
Catalog No. 502399145 Shop All Phenomenex Products
Encompass_Preferred
2mL centrifugal tube 150mgMgSO4, 50mgPSA, 20mgPestiCarb (GB23200.110-2018)
Catalog No. 50-239-9145 Supplier Phenomenex Inc Supplier No. MSPP0262
2mL centrifugal tube 150mgMgSO4, 50mgPSA, 20mgPestiCarb (GB23200.110-2018)