Learn More
Phenomenex Inc PSA 150mg, MgSO4 (Anhydrous) 150mg, C18 50mg
Catalog No. 502398400 Shop All Phenomenex Products
Encompass_Preferred
2mL Centrifugal Tube MS-9PA0206
Catalog No. 50-239-8400 Supplier Phenomenex Inc Supplier No. MS9PA0206
2mL Centrifugal Tube MS-9PA0206