Learn More
Phenomenex Inc 2mL centrifugal tube
Catalog No. 502399150 Shop All Phenomenex Products
Encompass_Preferred
2mL centrifugal tube 150mg MgSO4, 40mg PSA, 15mg PestiCarb(GB23200.110-2018)
Catalog No. 50-239-9150 Supplier Phenomenex Inc Supplier No. MSPP0263
2mL centrifugal tube 150mg MgSO4, 40mg PSA, 15mg PestiCarb(GB23200.110-2018)