Learn More
Phenomenex Inc 2mL Centrifugal Tube
Catalog No. 502400101 Shop All Phenomenex Products
Encompass_Preferred
2mL Centrifugal Tube PSA 75mg, C18 100mg
Catalog No. 50-240-0101 Supplier Phenomenex Inc Supplier No. MS9PA0229
2mL Centrifugal Tube PSA 75mg, C18 100mg