Learn More
Phenomenex Inc 2mL Centrifugal Tube
Catalog No. 502397979 Shop All Phenomenex Products
Encompass_Preferred
2mL Centrifugal Tube PSA 40mg, PC 10mg, MgSO4 (Anhydrous) 150mg
Catalog No. 50-239-7979 Supplier Phenomenex Inc Supplier No. MSPP0260
2mL Centrifugal Tube PSA 40mg, PC 10mg, MgSO4 (Anhydrous) 150mg