Learn More
Phenomenex Inc 2mL Centrifugal Tube
Catalog No. 502398258 Shop All Phenomenex Products
Encompass_Preferred
2mL Centrifugal Tube PSA 50mg, MgSO4 (Anhydrous) 100mg, C18 100mg
Catalog No. 50-239-8258 Supplier Phenomenex Inc Supplier No. MS9PA0205
2mL Centrifugal Tube PSA 50mg, MgSO4 (Anhydrous) 100mg, C18 100mg