Learn More
Andwin Scientific 2ML CENTRIFUGE TUBE
SDP
Catalog No. NC9829925
Encompass_Preferred
The disposable 2 mL microcentrifuge tubes are autoclavable, nonsterile, and graduated tubes that ensure easy and convenient sample measurement. This product also have flat caps and frosted areas on the sides that allows users easy sample identification and labeling. The caps can be opened with no difficulty, but will remain tightly closed during the process of centrifuging. Syringes can also penetrate the caps. This product is sold in the natural color.
Catalog No. NC9829925 Supplier Andwin Scientific Supplier No. 20170170
The disposable 2 mL microcentrifuge tubes are autoclavable, nonsterile, and graduated tubes that ensure easy and convenient sample measurement. This product also have flat caps and frosted areas on the sides that allows users easy sample identification and labeling. The caps can be opened with no difficulty, but will remain tightly closed during the process of centrifuging. Syringes can also penetrate the caps. This product is sold in the natural color.