Learn More
WELCH MATERIALS INC DoprahTM 2mL Amber Glass 12x32mm Flat Base 9-425 Screw Thread Vial with Label. Blue 9-425 Open Top Ribbed Screw Cap with 9mm Red PTFE/White Silicone Septa 1mm Thick (UltraClean). Blue Kit Packing.100pcs/pk
Catalog No. 502093130
Encompass_Preferred
UltraClean 2mL 9-425 Vial Kit: 2mL Amber Glass 12x32mm Flat Base 9-425 Screw Thread Vial with Label. Blue 9-425 Open Top Ribbed Screw Cap with 9mm Red PTFE/White Silicone Septa 1mm Thick (UltraClean). Blue Kit Packing.100pcs/pk
Catalog No. 50-209-3130 Supplier WELCH MATERIALS INC Supplier No. DS1172
UltraClean 2mL 9-425 Vial Kit: 2mL Amber Glass 12x32mm Flat Base 9-425 Screw Thread Vial with Label. Blue 9-425 Open Top Ribbed Screw Cap with 9mm Red PTFE/White Silicone Septa 1mm Thick (UltraClean). Blue Kit Packing.100pcs/pk