Learn More
WELCH MATERIALS INC DoprahTM 2mL Clear Glass 12x32mm Flat Base 9-425 Screw Thread Vial with Label. Blue 9-425 Open Top Ribbed Screw Cap with 9mm Red PTFE/White Silicone Septa 1mm Thick (UltraClean). Blue Kit Packing.100pcs/pk
Catalog No. 502093129
Encompass_Preferred
UltraClean 2mL 9-425 Vial Kit: 2mL Clear Glass 12x32mm Flat Base 9-425 Screw Thread Vial with Label. Blue 9-425 Open Top Ribbed Screw Cap with 9mm Red PTFE/White Silicone Septa 1mm Thick (UltraClean). Blue Kit Packing.100pcs/pk
Catalog No. 50-209-3129 Supplier WELCH MATERIALS INC Supplier No. DS1170
UltraClean 2mL 9-425 Vial Kit: 2mL Clear Glass 12x32mm Flat Base 9-425 Screw Thread Vial with Label. Blue 9-425 Open Top Ribbed Screw Cap with 9mm Red PTFE/White Silicone Septa 1mm Thick (UltraClean). Blue Kit Packing.100pcs/pk