Learn More
TWD Scientific LLC 2mL Internally Threaded Cryovial, Sterile. Case of 1000
SDP
Catalog No. NC0245057 Shop All TWD Scientific LLC Products
Encompass
STERILE 2mL (12.5x49mm) CLEAR POLYPROPYLENE (SELF-STANDING) CRYOVIALS WITH INTERNAL THREADS AND SILICONE WASHER SEAL CAPS ASSEMBLED. VIALS COME WITH WRITING PATCH AND MARKED GRADUATIONS. 1000/CASE
Catalog No. NC0245057 Supplier TWD Scientific LLC Supplier No. T3112
STERILE 2mL (12.5x49mm) CLEAR POLYPROPYLENE (SELF-STANDING) CRYOVIALS WITH INTERNAL THREADS AND SILICONE WASHER SEAL CAPS ASSEMBLED. VIALS COME WITH WRITING PATCH AND MARKED GRADUATIONS. 1000/CASE