Learn More
Analytical Sales & Services 2mL Clear Screw Vials and Caps - for Shimadzu Autosamplers
Catalog No. 5000804726
Encompass
Advantage™ Pre-Assembled Vial Kit. Includes: 8mm, 2mL, 12 X 32 Clear Glass Screw Vials and 8mm Black Caps with Soft Silicone/PTFE Septas For Shimadzu Autosamplers
Catalog No. 50-008-04726 Supplier Analytical Sales & Services Supplier No. 52008CASE
Advantage™ Pre-Assembled Vial Kit. Includes: 8mm, 2mL, 12 X 32 Clear Glass Screw Vials and 8mm Black Caps with Soft Silicone/PTFE Septas For Shimadzu Autosamplers