Learn More
Analytical Sales & Services 2mL Clear Screw Vials and Caps - for Spectra-Physics Autosamplers
Catalog No. 5000806953
Encompass
Advantage™ Pre-Assembled 8mm Vial Kit. Includes: 8mm, 2mL, 12 X 32 Clear Glass Screw Vials and 8mm Black Screw Caps with Silicone/PTFE Septas For Spectra-Physics Autosamplers
Catalog No. 50-008-06953 Supplier Analytical Sales & Services Supplier No. 52004CASE
Advantage™ Pre-Assembled 8mm Vial Kit. Includes: 8mm, 2mL, 12 X 32 Clear Glass Screw Vials and 8mm Black Screw Caps with Silicone/PTFE Septas For Spectra-Physics Autosamplers