Learn More
Gerstel 2ML CLEAR VIAL FOR MPS PK/100
SDP
Catalog No. NC9071353
Encompass
2 mL clear vials for MPS, Package of 100 Requires magnetic crimp caps with septa (093640-007-00)
Catalog No. NC9071353 Supplier Gerstel Supplier No. 09364000400
2 mL clear vials for MPS, Package of 100 Requires magnetic crimp caps with septa (093640-007-00)