Learn More
Analytical Sales & Services 2mL Clear-Vu™ 96-Well Collection Plate with Round-Bottom Wells
Catalog No. 5000804643
Encompass
Advantage™ Clear-Vu 2mL Collection Plate\n• Square Wells with Round Bottoms\n• High Quality, Solvent-Resistant See-Through Polypropylene\n• Industry Standard\n• Distinct Alphanumeric Designation\n• Compatible with All Robotic Systems\n\nQty/Pkg: 24
Catalog No. 50-008-04643 Supplier Analytical Sales & Services Supplier No. 27P557
Advantage™ Clear-Vu 2mL Collection Plate\n• Square Wells with Round Bottoms\n• High Quality, Solvent-Resistant See-Through Polypropylene\n• Industry Standard\n• Distinct Alphanumeric Designation\n• Compatible with All Robotic Systems\n\nQty/Pkg: 24