Learn More
Kent Laboratories Inc 2ML CONJ.C3 ANTI HUMAN
SDP
Catalog No. NC9842288
Encompass_Preferred
Fluorescein Conjugated Immunoglobulin Fractions,2ML,Anti Human Complement C-3 ,B1C/B1A,F/P Ratio:2-4.9,Monospecificity by solid phase adsorption for a strong tag with a low background,High antibody titer equals more dilution and dollar valu
Catalog No. NC9842288 Supplier Kent Laboratories Inc Supplier No. 105202
Fluorescein Conjugated Immunoglobulin Fractions,2ML,Anti Human Complement C-3 ,B1C/B1A,F/P Ratio:2-4.9,Monospecificity by solid phase adsorption for a strong tag with a low background,High antibody titer equals more dilution and dollar valu