Learn More
Waters Corp Cryogenic Storage Vial Domino, 2 mL, 24-Position, Andrew+ Compatible
Catalog No. NC3597548 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 2 mL Cryogenic Storage Vial Domino is engineered to securely hold up to 24 commonly used 2 mL cryogenic vials in individual positions. This product serves as a machine-readable consumable holder, establishing the crucial interface between various consumable types and the Andrew+ pipetting robot. This design facilitates seamless automation for a wide array of laboratory workflows.
  • Holds up to 24, 2 mL cryogenic vials in individual positions.
  • Designed for use with the Andrew+ pipetting robot.
  • Enables seamless automation of laboratory workflows.
  • Features QR codes for critical labware information.
  • Utilizes ARUCO codes for spatial positioning.
Catalog No. NC3597548 Supplier Waters Corp Supplier No. 186010163
The 2 mL Cryogenic Storage Vial Domino is engineered to securely hold up to 24 commonly used 2 mL cryogenic vials in individual positions. This product serves as a machine-readable consumable holder, establishing the crucial interface between various consumable types and the Andrew+ pipetting robot. This design facilitates seamless automation for a wide array of laboratory workflows.
  • Holds up to 24, 2 mL cryogenic vials in individual positions.
  • Designed for use with the Andrew+ pipetting robot.
  • Enables seamless automation of laboratory workflows.
  • Features QR codes for critical labware information.
  • Utilizes ARUCO codes for spatial positioning.