Learn More
Andwin Scientific 2.0 ml Dolphin MCT, Non-Sterile, Bulk Bag, 1,000 Tubes/Bag, 5 Bags/Case
SDP
Catalog No. NC9253992
Encompass_Preferred
The SafeSeal Microcentrifuge Tubes are compatible with most microcentrifuges and are ideal for biotechnology applications. The attached caps permit repeated openings and closings and also eliminate the risk of opening as a result of internal pressure or boiling. The tubes have dolphin bottom and have a volume capacity of 2 mL. Qty: 5 bags of 1,000 tubes each.
Catalog No. NC9253992 Supplier Andwin Scientific Supplier No. 16140
The SafeSeal Microcentrifuge Tubes are compatible with most microcentrifuges and are ideal for biotechnology applications. The attached caps permit repeated openings and closings and also eliminate the risk of opening as a result of internal pressure or boiling. The tubes have dolphin bottom and have a volume capacity of 2 mL. Qty: 5 bags of 1,000 tubes each.