Learn More
Andwin Scientific 2ML GRADUATED PIPETTES 6/PK
SDP
Catalog No. 50930567
Encompass_Preferred
The Electron Microscopy Science Calibrated Pipettes are constructed from translucent polypropylene, which make this product resistant to breakage. The pipettes feature blue graduations that permit users to precisely measure liquid samples. This product also has ring mark and inscriptions. These pipettes are ideal for use in fluoro and silicate chemistry procedures. This is sold in a pack of 6 pipettes with a volume of 2 mL.
Catalog No. 50-930-567 Supplier Andwin Scientific Supplier No. 7095102
The Electron Microscopy Science Calibrated Pipettes are constructed from translucent polypropylene, which make this product resistant to breakage. The pipettes feature blue graduations that permit users to precisely measure liquid samples. This product also has ring mark and inscriptions. These pipettes are ideal for use in fluoro and silicate chemistry procedures. This is sold in a pack of 6 pipettes with a volume of 2 mL.