Learn More
Sarstedt Inc 2ML LOOP SC MTUBE CBS 500/PK
Catalog No. NC1532999 Shop All Sarstedt Inc. Products
Encompass_Preferred
Screw Cap Micro Tube, 2 ml, PP, with skirted base, with knurls, with attached cap not screwed on, no print, 500 pcs./bag.
This item is not returnable. View return policy
Catalog No. NC1532999 Supplier Sarstedt Inc Supplier No. 72.694.100
Screw Cap Micro Tube, 2 ml, PP, with skirted base, with knurls, with attached cap not screwed on, no print, 500 pcs./bag.