Learn More
Boca Scientific Inc Magnetic Beads with silica surface, 3um bead size, 2ml
Supplier: Boca Scientific Inc MD41003
Magnetic beads are surface activated magnetic particles intended for covalent immobilization of proteins (e.g. antibodies, enzymes), peptides, nucleic acids or other molecules of interest. Different surface modifications and bead sizes allow for the selection of the optimal product for the application and molecule to be coupled. After the molecule of interest (ligand) is coupled to the magnetic particles, the resulting beads can be used in downstream applications such as detecting, enriching, or cleaving specific target molecules. The simple and efficient collection in magnetic fields allows for easy rinsing and removal of excess reagents and ligand after coupling. MD41003 beads consist of superparamagnetic silica particles and can be used for your own development purpose. They have a silica surface, a bead size of 3μm, and a concentration of 10mg/ml.