Learn More
Ace Glass, Inc. 2mL metal cap vial foil lined cap, cs/288
SDP
Catalog No. 50979875
Encompass
2mL sample vials fabricated from Wheaton-30 extractable glass. Include metal foil-lined caps. Vials reduce sample-to-surface contact when used as a micro package, and are economical when used as scintillation vials. Case/288.
Catalog No. 50-979-875 Supplier Ace Glass, Inc. Supplier No. 877902
2mL sample vials fabricated from Wheaton-30 extractable glass. Include metal foil-lined caps. Vials reduce sample-to-surface contact when used as a micro package, and are economical when used as scintillation vials. Case/288.