Learn More
Chemglass Life Sciences 2mL Micro Volumetric Flask, Class A, Tolerance: +-0.015mL
SDP
Catalog No. CG160302
Encompass_Preferred
2mL Micro Volumetric Flask, Class A, Tolerance: ¿0.015mL. Class ¿A¿ micro volumetric flask calibrated to contain. With 14/20 top outer joint and supplied complete with a CG-3000 14/20 hollow stopper. Flasks have hex base for stability and ¿V¿ shaped interior bottom for total sample retrieval.
Catalog No. CG160302 Supplier Chemglass Life Sciences Supplier No. CG160302
2mL Micro Volumetric Flask, Class A, Tolerance: ¿0.015mL. Class ¿A¿ micro volumetric flask calibrated to contain. With 14/20 top outer joint and supplied complete with a CG-3000 14/20 hollow stopper. Flasks have hex base for stability and ¿V¿ shaped interior bottom for total sample retrieval.