Learn More
MTC Bio Individually wrapped screw-cap microtubes, 2.0mL, with O-ring, , molded graduations, caps attached, sterile, 500/cs
Catalog No. 502268456
Encompass_Preferred
SureSeal™ sterile screw-cap microtubes come in a variety of sizes, cap configurations, graduation options, and now light-blocking amber. Made from ultra-clear polypropylene, these screw cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -90°C to +121°C. All SureSeal™ screw cap microtubes are sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000xg. Ultra-clear polypropylene. Molded or non-graduated options . Leak-proof O-ring seal. Light-blocking amber available. Sterile, RNase, DNase, Pyrogen free
This item is not returnable. View return policy
Catalog No. 50-226-8456 Supplier MTC Bio Supplier No. C2231W
SureSeal™ sterile screw-cap microtubes come in a variety of sizes, cap configurations, graduation options, and now light-blocking amber. Made from ultra-clear polypropylene, these screw cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -90°C to +121°C. All SureSeal™ screw cap microtubes are sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000xg. Ultra-clear polypropylene. Molded or non-graduated options . Leak-proof O-ring seal. Light-blocking amber available. Sterile, RNase, DNase, Pyrogen free