Learn More
S2 Media 2mL Methyl Red-Voges Proskauer (MR-VP) Broth in 15x105mm Tube, 50/box
Catalog No. 502995973
Encompass_Preferred
S2Media™ Methyl Red-Voges Proskauer (MR-VP) Broth is a differential medium used in qualitative procedures for the performance of the Methyl-Red (MR) and Voges-Proskauer (VP) tests. These tests are utilized in the identification of Gram-negative bacilli. • Product meets ASM and CLSI reference methods and performance criteria.• MR-VP Broth should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5973 Supplier S2 Media Supplier No. 5392
S2Media™ Methyl Red-Voges Proskauer (MR-VP) Broth is a differential medium used in qualitative procedures for the performance of the Methyl-Red (MR) and Voges-Proskauer (VP) tests. These tests are utilized in the identification of Gram-negative bacilli. • Product meets ASM and CLSI reference methods and performance criteria.• MR-VP Broth should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.