Learn More
Bellco Glass, Inc. 2ml Pasteur Pipette, Individually Wrapped. sterile 500/pk, 2000/cs
SDP
Catalog No. 502330130
Encompass_Preferred
2ml Pasteur Pipette, Individually Wrapped. sterile 500/pk, 2000/cs. Converntional consumables for quantitative transfer of liquids, in the field of experimental direction of pipetting operations such as biology and chemistry. Cell culture laboratory liquid tranfer, dosing, aspiration or other liquid transfer. • Material: medical grade transparent LDPE• Excellent transparency with clear graduations for easy observation• Good fluidity for easy control• High precision and drop repeatability• Sterilized by Ethylene Oxide
Catalog No. 50-233-0130 Supplier Bellco Glass, Inc. Supplier No. 451400301
2ml Pasteur Pipette, Individually Wrapped. sterile 500/pk, 2000/cs. Converntional consumables for quantitative transfer of liquids, in the field of experimental direction of pipetting operations such as biology and chemistry. Cell culture laboratory liquid tranfer, dosing, aspiration or other liquid transfer. • Material: medical grade transparent LDPE• Excellent transparency with clear graduations for easy observation• Good fluidity for easy control• High precision and drop repeatability• Sterilized by Ethylene Oxide