Learn More
USA Scientific Inc .2mL PCR tube,attd fl cap,nat
Catalog No. 502745485
Encompass
0.2 mL TempAssure PCR tube, attached frosted flat cap, natural. DNase, RNase, DNA, PCR inhibitor, and pyrogen free. 1000/bag.
Catalog No. 50-274-5485 Supplier USA Scientific Inc Supplier No. 14028100
0.2 mL TempAssure PCR tube, attached frosted flat cap, natural. DNase, RNase, DNA, PCR inhibitor, and pyrogen free. 1000/bag.