Learn More
Bellco Glass, Inc. 2mL Pipet Bulk Packed
SDP
Catalog No. 502329623
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets.Specifications Bidirectional positive and negative graduations are printed in bold black ink 10mL wide tip Serological pipet for viscous solutions. Wide tip is 4x larger than standard 10mL pipet 25 amp 50mL pipet features an antidrip tip for accurate delivery Printed indicator arrow for quickly locating negative graduations when using a pipet controller. Color Code Purple 30 Individually Wrapped Case Quantity 90
Catalog No. 50-232-9623 Supplier Bellco Glass, Inc. Supplier No. 229232000
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets.Specifications Bidirectional positive and negative graduations are printed in bold black ink 10mL wide tip Serological pipet for viscous solutions. Wide tip is 4x larger than standard 10mL pipet 25 amp 50mL pipet features an antidrip tip for accurate delivery Printed indicator arrow for quickly locating negative graduations when using a pipet controller. Color Code Purple 30 Individually Wrapped Case Quantity 90