Learn More
Bellco Glass, Inc. 2mL Pipette, MLA, Macro, Red
SDP
Catalog No. 502330513
Encompass_Preferred
2mL Pipette, MLA, Macro, RedSingle-stroke “to deliver” operationAvailable in 10 fixed volumes ranging from 1 mL- 10 mLAlso available as 1-5 mL Adjustable model with 0.1 mL incrementsAlso available as Selectable model for set volumes: 2 mL/3 mL/4 mL/5 mL All pipettes in stock for immediate shipmentFind tip size for your MLA pipette chartCase Quantity1Primary ColorRedPrimary MaterialAluminum Alloy & Stainless SteelVolume2mLVistaLab Tip Fit SizeMacro 5Universal FitNoAccuracy±0.75%Precision0.50%
Catalog No. 50-233-0513 Supplier Bellco Glass, Inc. Supplier No. 451900081
2mL Pipette, MLA, Macro, RedSingle-stroke “to deliver” operationAvailable in 10 fixed volumes ranging from 1 mL- 10 mLAlso available as 1-5 mL Adjustable model with 0.1 mL incrementsAlso available as Selectable model for set volumes: 2 mL/3 mL/4 mL/5 mL All pipettes in stock for immediate shipmentFind tip size for your MLA pipette chartCase Quantity1Primary ColorRedPrimary MaterialAluminum Alloy & Stainless SteelVolume2mLVistaLab Tip Fit SizeMacro 5Universal FitNoAccuracy±0.75%Precision0.50%