Learn More
HEPATOCHEM 2 ml quartz vial
Catalog No. NC3245553
Encompass_Preferred
2 ml quartz vials 12x32mm with 10-425 thread (cap with septum included)
Catalog No. NC3245553 Supplier HEPATOCHEM Supplier No. HCK102101051
2 ml quartz vials 12x32mm with 10-425 thread (cap with septum included)