Learn More
Sarstedt Inc 2mL Screw Cap Micro Tube, Conical Bottom, Sterile, 1000/Case
Catalog No. 50809245 Shop All Sarstedt Inc. Products
Encompass_Preferred
2mL Screw Cap Micro Tube Conical Bottom Skirted Graduations and Writing Block Mixed Color O-Ring Caps Sterile
This item is not returnable. View return policy
Catalog No. 50-809-245 Supplier Sarstedt Inc Supplier No. 72.694.996
2mL Screw Cap Micro Tube Conical Bottom Skirted Graduations and Writing Block Mixed Color O-Ring Caps Sterile