Learn More
Andwin Scientific 2ML SCREW CAP MICRO TUBES NAT.
SDP
Catalog No. NC9031476
Encompass_Preferred
The SuperClear™ Screw Cap Microcentrifuge Tubes feature an ultra-clear resin that permits easy viewing of contents. This product has universal screw threads that are compatible with most popular brand screw caps. The tubes are freestanding and nonste
Catalog No. NC9031476 Supplier Andwin Scientific Supplier No. 16466060
The SuperClear™ Screw Cap Microcentrifuge Tubes feature an ultra-clear resin that permits easy viewing of contents. This product has universal screw threads that are compatible with most popular brand screw caps. The tubes are freestanding and nonste