Learn More
Bellco Glass, Inc. 2mL Serological Pipet Individually Wrapped
SDP
Catalog No. 502329775
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identificationPrinted indicator arrow for quickly locating negative graduations when using a pipet controllerPack Description100 Individually Wrapped/BagCase Quantity500SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorGreenPrimary MaterialPolystyreneMaximum Volume2mLGraduatedYesGraduation Increments (Subdivisions)1/100mLRNase/DNase FreeYes
Catalog No. 50-232-9775 Supplier Bellco Glass, Inc. Supplier No. 229002B00
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identificationPrinted indicator arrow for quickly locating negative graduations when using a pipet controllerPack Description100 Individually Wrapped/BagCase Quantity500SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorGreenPrimary MaterialPolystyreneMaximum Volume2mLGraduatedYesGraduation Increments (Subdivisions)1/100mLRNase/DNase FreeYes