Learn More
CELLTREAT Scientific Products 2mL Serological Pipet, Individual Paper/Plastic Wrapper Packed in Bags, Sterile pipette
SDP
Catalog No. 50202104
Encompass_Preferred
CELLTREAT Scientific Products 2mL Serological Pipet. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in paper/plastic wrappers. 100 packed/bag, 600/case.
Catalog No. 50-202-104 Supplier CELLTREAT Scientific Products Supplier No. 229202B
CELLTREAT Scientific Products 2mL Serological Pipet. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in paper/plastic wrappers. 100 packed/bag, 600/case.