Learn More
CELLTREAT Scientific Products 2mL Serological Pipet, Bulk Packed in Bags, Sterile pipette
SDP
Catalog No. 50202121
Encompass_Preferred
CELLTREAT Scientific Products 2mL Serological Pipet. Convenient bulk packaging option. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase free and non-pyrogenic. -0.5mL graduations. Every case is printed with the lot number for quality traceability. Packaged sterile, 25 bulk packed/re-sealable bag, 800/case.
Catalog No. 50-202-121 Supplier CELLTREAT Scientific Products Supplier No. 229232
CELLTREAT Scientific Products 2mL Serological Pipet. Convenient bulk packaging option. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase free and non-pyrogenic. -0.5mL graduations. Every case is printed with the lot number for quality traceability. Packaged sterile, 25 bulk packed/re-sealable bag, 800/case.