Learn More
Ace Glass, Inc. 2ML SPECIFIC GRAVITY BOTTLE
SDP
Catalog No. 50971880
Encompass_Preferred
Guy-Lussac type 2mL specific gravity bottle, unadjusted for calibration in the laboratory. When adjusted, suitable for ASTM D 369.
Catalog No. 50-971-880 Supplier Ace Glass, Inc. Supplier No. 542004
Guy-Lussac type 2mL specific gravity bottle, unadjusted for calibration in the laboratory. When adjusted, suitable for ASTM D 369.