Learn More
USA Scientific Inc .2mL TempAssure PCR 8-tube
Catalog No. 502744729
Encompass
0.2 mL TempAssure PCR 8-tube flex-free strips with individually attached, optically clear flat caps. Tubes are assorted colors, caps are natural. 10 bags of 12 strips.
Catalog No. 50-274-4729 Supplier USA Scientific Inc Supplier No. 14024708
0.2 mL TempAssure PCR 8-tube flex-free strips with individually attached, optically clear flat caps. Tubes are assorted colors, caps are natural. 10 bags of 12 strips.