Learn More
USA Scientific Inc .2mL TempAssure PCR white
Catalog No. 502745327
Encompass
0.2 mL TempAssure PCR white 8-tube strips with separate optically clear 8-cap (flat top) strips. 125 strips each of tubes and caps per pack.
Catalog No. 50-274-5327 Supplier USA Scientific Inc Supplier No. 14022580
0.2 mL TempAssure PCR white 8-tube strips with separate optically clear 8-cap (flat top) strips. 125 strips each of tubes and caps per pack.