Learn More
Analytical Sales & Services 2mL TrueTaper 96-Well Collection Plate with 100uL Tapered-Bottom Wells, Irradiated
Catalog No. 5000804213
Encompass
2mL TrueTaper 96-well Collection Plate, Irradiated\n\n• Unique Tapered Bottom for Maximum Sample Recovery\n• Square Wells with 100µL TAPERED Bottoms (2mL Total Volume per Well)\n• Irradiated\n• Made in USA of domestically sourced and certified materials\n\nQty/Pkg: 25
Catalog No. 50-008-04213 Supplier Analytical Sales & Services Supplier No. 968820RAD
2mL TrueTaper 96-well Collection Plate, Irradiated\n\n• Unique Tapered Bottom for Maximum Sample Recovery\n• Square Wells with 100µL TAPERED Bottoms (2mL Total Volume per Well)\n• Irradiated\n• Made in USA of domestically sourced and certified materials\n\nQty/Pkg: 25