Learn More
Analytical Sales & Services 2mL TrueTaper 96-Well Collection Plate with 100uL Tapered-Bottom Wells, with SiliGuard™ Low Bind Coating, Irradiated
Catalog No. 5000804585
Encompass
2mL TrueTaper 96-well Collection Plate with SiliGuard™, Irradiated\n\n• Unique Tapered Bottom for Maximum Sample Recovery\n• Square Wells with 100µL TAPERED Bottoms (2mL Total Volume per Well)\n• SiliGuard™ Low Bind Coating\n• Irradiated\n• Made in USA of domestically sourced and certified materials\n\nQty/Pkg: 25
Catalog No. 50-008-04585 Supplier Analytical Sales & Services Supplier No. 968820GCRAD
2mL TrueTaper 96-well Collection Plate with SiliGuard™, Irradiated\n\n• Unique Tapered Bottom for Maximum Sample Recovery\n• Square Wells with 100µL TAPERED Bottoms (2mL Total Volume per Well)\n• SiliGuard™ Low Bind Coating\n• Irradiated\n• Made in USA of domestically sourced and certified materials\n\nQty/Pkg: 25