Learn More
BioSpec Products 2ML VIAL WITH CAP, 500/PK
SDP
Catalog No. NC0461857
Encompass_Preferred
Our 2.0 ml, screw cap with o-ring seal, microcentrifuge vials are made of inert polypropylene and have straight walls with a sharp conical bottom.
Catalog No. NC0461857 Supplier BioSpec Products Supplier No. 10832
Our 2.0 ml, screw cap with o-ring seal, microcentrifuge vials are made of inert polypropylene and have straight walls with a sharp conical bottom.