Learn More
Genesee Scientific Corporation Olympus 2.0ml Microtubes, Clear, No Cap
SDP
Catalog No. NC1433170
Encompass
Olympus 2.0ml capless microcentrifuge tubes are identical to Olympus's capped version (cat# 22-283) in every way except for the lack of a cap. They are ready for autoclaving at 121ºC for 15 minutes. The tubes are exceptionally clear and can be used in any centrifuge up to 20,000 x g.
Catalog No. NC1433170 Supplier Genesee Scientific Corporation Supplier No. 22283NC
Olympus 2.0ml capless microcentrifuge tubes are identical to Olympus's capped version (cat# 22-283) in every way except for the lack of a cap. They are ready for autoclaving at 121ºC for 15 minutes. The tubes are exceptionally clear and can be used in any centrifuge up to 20,000 x g.