Learn More
Arcticwhite LLC 96 Square Well, 2 ML/Well, Polypropylene, Round-Conical Bottom, Natural, Non-sterilized, 96 Wells, SBS Footprint, Medical-grade Polypropylene (PP), 25 Microplates per Case
SDP
Catalog No. NC9385014
Encompass
96 Square Well, 2 ML, Polypropylene, Round-Conical Bottom, Natural, Non-sterilized, 96 Wells, SBS Footprint, 2 ML per Well, Medical-grade Polypropylene (PP), 25 Microplates per Case. This deep-well microplate is used for automated liquid handling, nucleic acid purification, and cell or bacterial culture. Format - 96 wells, SBS footprint. Well shape - square. Bottom - round-conical. Well volume - 2 mL per well. Material - medical-grade polypropylene (PP). Certifications - SBS standard. Supplied by ArcticWhite as 25 microplates per case.
Catalog No. NC9385014 Supplier Arcticwhite LLC Supplier No. AWLSX10006
96 Square Well, 2 ML, Polypropylene, Round-Conical Bottom, Natural, Non-sterilized, 96 Wells, SBS Footprint, 2 ML per Well, Medical-grade Polypropylene (PP), 25 Microplates per Case. This deep-well microplate is used for automated liquid handling, nucleic acid purification, and cell or bacterial culture. Format - 96 wells, SBS footprint. Well shape - square. Bottom - round-conical. Well volume - 2 mL per well. Material - medical-grade polypropylene (PP). Certifications - SBS standard. Supplied by ArcticWhite as 25 microplates per case.