Learn More
Chemglass Life Sciences Stopcock, Four Way, PTFE Plug, 2mm Bore, 15.2/30 Plug Size, 8mm OD Sidearms
SDP
Catalog No. CG44301
Encompass_Preferred
Stopcock, Four Way, PTFE Plug, 2mm Bore, 15.2/30 Plug Size, 8mm OD Sidearms. A 1:5 taper PTFE plug having double "V", 90º bores to connect opposite sets of adjacent arms. Configuration of stopcock allows it to be used in a system to obtain reversal of flow.
Catalog No. CG44301 Supplier Chemglass Life Sciences Supplier No. CG44301
Stopcock, Four Way, PTFE Plug, 2mm Bore, 15.2/30 Plug Size, 8mm OD Sidearms. A 1:5 taper PTFE plug having double "V", 90º bores to connect opposite sets of adjacent arms. Configuration of stopcock allows it to be used in a system to obtain reversal of flow.